CAS RN 54460-46-7
CAS RN 54460-46-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cycloprate, TRC
CAS: 54460-46-7 Molecular Formula: C20 H38 O2 Molecular Weight (g/mol): 310.51 Synonym: Hexadecyl Cyclopropanecarboxylate,ZR 856,Zardex,Cyclopropanecarboxylic Acid, Hexadecyl Ester; IUPAC Name: hexadecyl cyclopropanecarboxylate SMILES: CCCCCCCCCCCCCCCCOC(=O)C1CC1