missing translation for 'onlineSavingsMsg'
Learn More

Cyclophilin-E Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

190.00€ - 550.00€

Specifications

Antigen Cyclophilin-E
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30230488
View Documents
Novus Biologicals
NBP3-38374-20ul
20 μL
190.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30229303
View Documents
Novus Biologicals
NBP3-38374-100ul
100 μL
550.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclophilin-E Polyclonal antibody specifically detects Cyclophilin-E in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications

Specifications

Cyclophilin-E
ELISA, Immunoprecipitation, Western Blot
Unconjugated
Rabbit
DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes
PBS (pH 7.3), 50% glycerol
10450
IgG
Affinity purified
Western Blot 1:500 - 1:2000, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells
Polyclonal
Purified
RUO
Human, Mouse
Cyclophilin E, Cyclophilin-33, CyP-33, CYP33, EC 5.2.1.8, MGC111222, MGC3736, peptidyl-prolyl cis-trans isomerase E, peptidylprolyl isomerase E (cyclophilin E), PPIase E, Rotamase E
Recombinant fusion protein containing a sequence corresponding to amino acids 1-301 of human Cyclophilin-E (NP_006103.1).,, Sequence:, MATTKRVLYVGGLAEEVDDKVLHAAFIPFGDITDIQIPLDYETEKHRGFAFVEFELAEDAAAAIDNMNESELFGRTIRVNLAKPMRIKEGSSRPVWSDDDWLKKFSGKTLEENKEEEGSEPPKAETQEGEPIAKKARSNPQVYMDIKIGNKPAGRIQMLLRSDVVPMTAENFRCLCTHEKGFGFKGSSFHRIIPQFMCQGGDFTNHNGTGGKSIYGKKFDDENFILKHTGPGLLSMANSGPNTNGSQFFLTCDKTDWLDGKHVVFGEVTEGLDVLRQIEAQGSKDGKPKQKVIIADCGEYV
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.