missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cyclophilin B Polyclonal specifically detects Cyclophilin B in Human, Mouse, Porcine, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Knockout Validated.
Specifications
Specifications
| Antigen | Cyclophilin B |
| Applications | Western Blot, KnockDown |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-2 ug/ml, Knockout Validated |
| Formulation | 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Accession No. | P23284 |
| Gene Alias | Cyclophilin B, cyclophilin-like protein, CYPBEC 5.2.1.8, CYP-S1, MGC2224, OI9MGC14109, peptidyl-prolyl cis-trans isomerase B, peptidylprolyl isomerase B (cyclophilin B), PPIase B, Rotamase B, S-cyclophilin, SCYLP |
| Gene Symbols | PPIB |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?