Learn More
Cycloate, Dr. Ehrenstorfer
Product Code. 30311836
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311836 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30311836 Supplier Dr. Ehrenstorfer Supplier No. DREC11820000
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cycloate
CAS 1134-23-2
Analyte or Component Names Cycloate
Shipping Condition Room Temperature
Molecular Formula C11 H21 N O S
Certifications/Compliance ISO 17034
Synonym Carbamothioic acid, cyclohexylethyl-, S-ethyl ester (9CI), Cyclohexanecarbamic acid, N-ethylthio-, S-ethyl ester (7CI,8CI), Buranit, Cycloate, Ethyl N-cyclohexyl-N-ethylthiolcarbamate, Etsan, Eurex, Hexylthiocarbam, R 2063, Ro-Neet, Ro-Neet 6E, Roneet 72LE, Ronit, S-Ethyl N-cyclohexyl-N-ethylthiocarbamate, S-Ethyl N-cyclohexyl-N-ethylthiolcarbamate, S-Ethyl cyclohexylethylthiocarbamate, S-Ethyl-N-ethyl-N-cyclohexylthiolcarbamate, Sabet
InChI Formula InChI=1S/C11H21NOS/c1-3-12(11(13)14-4-2)10-8-6-5-7-9-10/h10H,3-9H2,1-2H3
Solution Type Neat
SMILES CCSC(=O)N(CC)C1CCCCC1
IUPAC Name S-ethyl N-cyclohexyl-N-ethylcarbamothioate
Industry Type Environmental Testing
Molecular Weight (g/mol) 215.36
Quantity 250 mg
Formula Weight 215.1344 g/mol