missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin H Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33310-100ul
This item is not returnable.
View return policy
Description
Cyclin H Monoclonal antibody specifically detects Cyclin H in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Cyclin H | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| CAK, cyclin H, cyclin-dependent kinase-activating kinase complex subunit, cyclin-H, MO15-associated protein, p34CAK complex subunit, p37CDK-activating kinase complex subunit | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Cyclin H (P51946).,, Sequence:, MYHNSSQKRHWTFSSEEQLARLRADANRKFRCKAVANGKVLPNDPVFLEPHEEMTLCKYYEKRLLEFCSVFKPAMPRSVVGTACMYFKRFYLNNSVMEYH | |
| 100 μL | |
| Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair, Phospho Specific | |
| 902 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction