missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35328-100ul
This item is not returnable.
View return policy
Description
Cyclin D2 Polyclonal antibody specifically detects Cyclin D2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Cyclin D2 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| cyclin D2, G1/S-specific cyclin D2, G1/S-specific cyclin-D2, KIAK0002, MGC102758 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-289 of human Cyclin D2 (NP_001750.1).,, Sequence:, MELLCHEVDPVRRAVRDRNLLRDDRVLQNLLTIEERYLPQCSYFKCVQKDIQPYMRRMVATWMLEVCEEQKCEEEVFPLAMNYLDRFLAGVPTPKSHLQLLGAVCMFLASKLKETSPLTAEKLCIYTDNSIKPQELLEWELVVLGKLKWNLAAVTPHDFIEHILRKLPQQREKLSLIRKHAQTFIALCATDFKFAMYPPSMIATGSVGAAICGLQQDEEVSSLTCDALTELLAKITNTDVDCLKACQEQIEAVLLNSLQQYRQDQRDGSKSEDELDQASTPTDVRDIDL | |
| 100 μL | |
| Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Wnt Signaling Pathway | |
| 894 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction