missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D1 Antibody (rCCND1/4752) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08953
This item is not returnable.
View return policy
Description
Cyclin D1 Monoclonal specifically detects Cyclin D1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| Cyclin D1 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 ug/mL | |
| B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 | |
| Synthetic peptide corresponding to residues within aa 200-295 of Cyclin D1 | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
| 595 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| rCCND1/4752 | |
| Unconjugated | |
| 10mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction