missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D1 Antibody (CL12328) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21214JF525
This item is not returnable.
View return policy
Description
Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Cyclin D1 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 | |
| This antibody was developed using a synthetic peptide derived from P24385, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
| 595 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunocytochemistry/Immunofluorescence | |
| CL12328 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction