missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67726-1-IG-150UL
This item is not returnable.
View return policy
Description
The Cyclin B2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cyclin B2. This antibody recognizes human, mouse antigen. The Cyclin B2 antibody has been validated for the following applications: WB, IHC, IF/ICC, IF-P, ELISA analysis.Specifications
| Cyclin B2 | |
| Monoclonal | |
| 1600 μg/mL | |
| HsT17299 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| 45 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, ELISA | |
| 2G8B7 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 9133 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu