Learn More
Novus Biologicals™ CYB5R3 Protein
Product Code. p-200039045 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100μg
Unit Size:
0.50mg
100µg
Product Code. 18466290
Brand: Novus Biologicals™ NBP1788790.5mg
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 27-301 of Human CYB5R3 The Recombinant Human CYB5R3 Protein is derived from E. coli. The Recombinant Human CYB5R3 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20 mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 1 mM DTT, 0.1 M NaCl.
Molecular Weight (g/mol) 34 kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Human
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name CYB5R3
Conjugate Unconjugated
Purity or Quality Grade >95%