missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR4 Antibody (ulocuplumab) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28275JF669
This item is not returnable.
View return policy
Description
CXCR4 Monoclonal antibody specifically detects CXCR4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CXCR4 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor | |
| CXCR4/ CD184 | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Metastasis, Neuronal Cell Markers, Signal Transduction, Stem Cells | |
| 7852 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| ulocuplumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction