missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR1/IL-8RA Antibody (8F1-1-4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
252.00€ - 451.00€
Specifications
| Antigen | CXCR1/IL-8 RA |
|---|---|
| Clone | 8F1-1-4 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Fluorophore-linked immunosorbent assay 1:100-1:2000 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18222493
|
Novus Biologicals
NBP1-43339-0.025MG |
0.025 mg |
252.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18238774
|
Novus Biologicals
NBP1-43339-0.1MG |
0.1 mg |
451.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR1/IL-8RA Monoclonal specifically detects CXCR1/IL-8RA in Human, Hamster samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Fluorophore-linked immunosorbent assay.Specifications
| CXCR1/IL-8 RA | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| P25024 | |
| 3577 | |
| The immunogen for this antibody was CD181. | |
| Primary | |
| Store at 4C. Do not freeze. |
| 8F1-1-4 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Fluorophore-linked immunosorbent assay 1:100-1:2000 | |
| Monoclonal | |
| Mouse | |
| Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A | |
| CXCR1 | |
| IgG2b κ | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title