missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL10/IP-10/CRG-2 Antibody (5C7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26638-50ul
This item is not returnable.
View return policy
Description
CXCL10/IP-10/CRG-2 Monoclonal antibody specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| CXCL10/IP-10/CRG-2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 5C7 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| C7, chemokine (C-X-C motif) ligand 10, crg-2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
| A synthesized peptide derived from Human CXCL10/IP-10/CRG-2 [UniProt P02778] | |
| 50 μL | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 3627 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto