missing translation for 'onlineSavingsMsg'
Learn More

CXCL10/IP-10/CRG-2 Antibody (5C7), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-26638-100ul

459.00 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


 View more versions of this product

Product Code. 30043473

  • 533.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CXCL10/IP-10/CRG-2 Monoclonal antibody specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CXCL10/IP-10/CRG-2
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
5C7
ELISA, Immunohistochemistry 1:50-1:200
C7, chemokine (C-X-C motif) ligand 10, crg-2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
A synthesized peptide derived from Human CXCL10/IP-10/CRG-2 [UniProt P02778]
100 μL
Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
3627
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.