missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CWC22 Polyclonal antibody specifically detects CWC22 in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CWC22 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000 |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | CWC22 spliceosome-associated protein homolog (S. cerevisiae), EIF4GL, fSAPbpre-mRNA-splicing factor CWC22 homolog, KIAA1604NCMfunctional spliceosome-associated protein b, Nucampholin homolog |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 639-908 of human CWC22 (NP_065994.1). LTDELREHLKNTPKVIVAQKPDVEQNKSSPSSSSSASSSSESDSSDSDSDSSDSSSESSSEESDSSSISSHSSASANDVRKKGHGKTRSKEVDKLIRNQQTNDRKQKERRQEHGHQETRTERERRSEKHRDQNSSGSNWRDPITKYTSDKDVPSERNNYSRVANDRDQEMHIDLENKHGDPKKKRGERRNSFSENEKHTHRIKDSENFRRKDRSKSKEMNRKHSGSRSDEDRYQNGAERRWEKSSRYSEQSRESKKNQDRRREKSPAKQK |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?