missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CUX1 Polyclonal Antibody

Product Code. 30534690
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30534690 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534690 Supplier Invitrogen™ Supplier No. PA5148895

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUX1
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene CUX1
Gene Accession No. P39880, Q13948
Gene Alias CASP; CCAAT displacement protein; CDP; CDP/Cut; CDP1; CDP2; Cdpl1; Clox; COY1; cut homolog; cut like homeobox 1; CUTL1; cut-like 1; cut-like homeobox 1; Cux; Cux/CDP; Cux/CDP homeoprotein; CUX1; Cux-1; golgi integral membrane protein 6; GOLIM6; Homeobox protein cut-like 1; homeobox protein cux-1; Kiaa4047; Nbla10317; p100; p110; p200; p75; Protein CASP; putative protein product of Nbla10317
Gene Symbols CUX1
Host Species Goat
Immunogen E. coli-derived recombinant human CDP/CUTL1, Ile64-Ser268.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1523
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.