missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CUTL2 Monoclonal antibody specifically detects CUTL2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | CUTL2 |
| Applications | Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | CL14332 |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | CDP2, CUTL2, cut-like 2 (Drosophila), cut-like homeobox 2, homeobox protein cut-like 2, Homeobox protein cux-2, KIAA0293 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from O14529, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?