Learn More
R&D Systems™ Cultrex Poly-L-Ornithine Solution
Product Code. 18777696
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18777696 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18777696 Supplier R&D Systems™ Supplier No. 343610001
  • ONLINE EXCLUSIVE PRICE
    170.00€
    160.65€ / 100mL
    Save 9.35€
    5% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Edge
Add to basket
R&D Systems™ Cultrex™ Poly-L-Ornithine Solution is a highly positively charged amino acid chain, which is generally used as a coating reagent to promote cell adhesion in culture.
  • Poly-L-Ornithine along or in combination with Fibronectin or Laminin-I is frequently used to enhance attachment and differentiation of various types of neuronal and neural stem cells.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
Storage Requirements Product is stable for at least 6 months from the date of receipt when stored at 2 - 8°C. Keep sterile.
Species Multi-species
Product Line R&D Systems