missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex Mouse Laminin I, Pathclear

Product Code. 18790509
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18790509

Brand: R&D Systems™ 340001002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Bioactivity

Functional Assays: Supports the attachment of MG63 osteosarcoma cells at < 20 µg/ml.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation 1 mg/mL at purity of =90% (SDS-Page). Storage Buffer: Dulbecco's Modified Eagle's Medium (DMEM) with 10 μg/mL gentamicin sulfate.
Gene ID (Entrez) 284217
Quantity 1 mg
Storage Requirements Product is stable for a minimum of 3 months from date of shipment when stored at -20°C in a manual defrost freezer. For optimal stability, store at -80°C. Avoid freeze-thaw cycles.
Endotoxin Concentration <20.0 EU per 1μg of the protein by the LAL method.
Gene Alias LAMA, LAMA1, laminin subunit alpha 1, Laminin1, Laminin-111, PTBHS, S-LAM-alpha
Species Mouse
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.