Learn More
Cullin 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-58788
This item is not returnable.
View return policy
Description
Cullin 3 Polyclonal specifically detects Cullin 3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| 88.93 kDa | |
| IgG |
| CUL-3, cullin 3, cullin-3, KIAA0617 | |
| Store at 4C. Do not freeze. |