Learn More
Cullin 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NB100-58788
Product Code. 18795493
  • 595.00€ / 0.10mL
Description
Cullin 3 Polyclonal specifically detects Cullin 3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
88.93 kDa
IgG
CUL-3, cullin 3, cullin-3, KIAA0617
Store at 4C. Do not freeze.