missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTPS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12852-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene catalyzes the formation of CTP from UTP with the concomitant deamination of glutamine to glutamate. This protein is the rate-limiting enzyme in the synthesis of cytosine nucleotides, which play an important role in various metabolic processes and provide the precursors necessary for the synthesis of RNA and DNA. Cancer cells that exhibit increased cell proliferation also exhibit an increased activity of this encoded protein. Thus, this protein is an attractive target for selective chemotherapy. Alternative splicing results in multiple transcript variants.Specifications
| CTPS2 | |
| Polyclonal | |
| Unconjugated | |
| CTPS2 | |
| CTP synthase 2, CTP synthase II, CTP synthetase 2, CTPS2, UTP ammonia ligase 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55936, 56474, 619580 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P70303, Q5U2N0, Q9NRF8 | |
| Ctps2 | |
| CTPS2 Fusion Protein Ag3490 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction