Learn More
CTP synthase Antibody (13B6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26640-100ul
This item is not returnable.
View return policy
Description
CTP synthase Monoclonal antibody specifically detects CTP synthase in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| CTP synthase | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 13B6 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| CTP synthase, CTP synthase 1, CTP synthetase 1, cytidine 5-prime triphosphate synthetase, cytidine 5'-triphosphate synthetase, EC 6.3.4.2, UTP--ammonia ligase 1 | |
| A synthesized peptide derived from Human CTP synthase [UniProt P17812] | |
| 100 μL | |
| Endocrinology, Signal Transduction | |
| 1503 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |