Learn More
CTP synthase Antibody (13B6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26640-50ul
 View more versions of this product
Product Code. 30043457
  • 343.00€ / 50µL
Description
CTP synthase Monoclonal antibody specifically detects CTP synthase in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CTP synthase
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
13B6
ELISA, Immunohistochemistry 1:50-1:200
CTP synthase, CTP synthase 1, CTP synthetase 1, cytidine 5-prime triphosphate synthetase, cytidine 5'-triphosphate synthetase, EC 6.3.4.2, UTP--ammonia ligase 1
A synthesized peptide derived from Human CTP synthase [UniProt P17812]
50 μL
Endocrinology, Signal Transduction
1503
Store at -20 to -70C. Avoid freeze-thaw cycles.