missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The CTCF antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CTCF. This antibody recognizes human, mouse antigen. The CTCF antibody has been validated for the following applications: WB, IHC, IF/ICC, IP, ELISA analysis.
Specifications
Specifications
| Antigen | CTCF |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA |
| Classification | Polyclonal |
| Concentration | 700 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Recombinant Protein |
| Purification Method | Antigen affinity purification |
| Quantity | 20 μL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?