missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CT83 Antibody (CL4762) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59062UV
This item is not returnable.
View return policy
Description
CT83 Monoclonal antibody specifically detects CT83 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| CT83 | |
| Monoclonal | |
| DyLight 350 | |
| chromosome X open reading frame 61, CT83, CT83Cancer/testis antigen 83, CXorf61, FLJ20611, FLJ22913, kita-kyushu lung cancer antigen 1, KKLC1, KK-LC-1, KK-LC-1KKLC1, RP3-452H17.2 | |
| This antibody was developed using a recombinant protein derived from Q5H943, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL4762 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 203413 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction