missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CT47A11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55513-25ul
This item is not returnable.
View return policy
Description
CT47A11 Polyclonal specifically detects CT47A11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CT47A11 | |
| Polyclonal | |
| Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| Cancer/testis antigen 47, cancer/testis antigen 47A, cancer/testis antigen family 47, member A10, cancer/testis CT47 family, member 10, CT47, CT47.1, CT47.10CT47.3, CT47.11, CT47.12, CT47.2, CT47.4, CT47.5, CT47.6, CT47.7, CT47.8, CT47.9 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 255313 | |
| Human | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| CT47A11 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:PEEVTKSQPEKWDEEAQDAAGEEEKEQE | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction