missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSRP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Specifications
| Antigen | CSRP1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 μL/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601162
|
Novus Biologicals
NBP2-98292-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682612
|
Novus Biologicals
NBP2-98292-50ul |
50 μg |
216.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CSRP1 Polyclonal antibody specifically detects CSRP1 in Mouse samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| CSRP1 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Stem Cells | |
| 0.2 um filtered solution in PBS | |
| 1465 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 μL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CRP, CRP1, CSRPCYRP, cysteine and glycine-rich protein 1, Cysteine-rich protein 1, D1S181E, DKFZp686M148, LIM-domain protein | |
| Produced in rabbits immunized with purified, recombinant Mouse CSRP1 (Uniprot#: P97315; Met1-Glu193) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title