missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSHL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Specifications
| Antigen | CSHL1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18606235
|
Novus Biologicals
NBP2-98959-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659615
|
Novus Biologicals
NBP2-98959-50ul |
50 μg |
216.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CSHL1 Polyclonal antibody specifically detects CSHL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CSHL1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| chorionic somatomammotropin hormone-like 1, Chorionic somatomammotropin-like, CS-5, CSHP1, CSLchorionic somatomammotropin CS-5, growth hormone cluster, hCS-L, Lactogen-like, MGC149868 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CSHL1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| 1444 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title