missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSF3R Recombinant Human Monoclonal Antibody (11C11)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560775
This item is not returnable.
View return policy
Description
CSF3R Recombinant Monoclonal Antibody for IHC (P), ELISA
CSF3R is the receptor for colony stimulating factor 3, a cytokine that controls the production, differentiation, and function of granulocytes. The encoded protein, which is a member of the family of cytokine receptors, may also function in some cell surface adhesion or recognition processes. Four transcript variants encoding four different isoforms have been found for this gene, with three of the isoforms being membrane-bound and the other being secreted and soluble.
Specifications
| CSF3R | |
| Recombinant Monoclonal | |
| 1.24 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q99062 | |
| CSF3R | |
| Recombinant Human CSF3R protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin) | |
| 11C11 | |
| Unconjugated | |
| CSF3R | |
| CD114; CD114 antigen; colony stimulating factor 3 receptor; colony stimulating factor 3 receptor (granulocyte); Csf3r; Csfgr; G-CSF receptor; GCSFR; G-CSFR; G-CSF-R; Granulocyte colony-stimulating factor receptor; SCN7 | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 1441 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction