Learn More
CS Citrate Synthase Antibody (CL2553) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-36773PCP
Product Code. 30298181
  • 737.00€ / 0.10mL
Description
CS Citrate Synthase Monoclonal antibody specifically detects CS Citrate Synthase in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CS Citrate Synthase
Monoclonal
PerCP
citrate synthase, citrate synthase, mitochondrial, EC 2.3.3, EC 2.3.3.1
This antibody was developed using a recombinant protein derived from O75390, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Endocrinology, Signal Transduction
1431
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL2553
PBS
Mouse
Protein A purified