Description
- Part of Eppendorf CryoStorage Vials
Specifications
Specifications
| Color | Black |
| Quantity | 10 Pk. |
| Material | Polypropylene |
| Type | Cryogenic Vial Rack |
| Capacity (Metric) | 0.5 mL |
| Withstand Temperature (Metric) | -80°C |