Learn More
Cryoscope Standard 621 (-0.600°C) (621m°H), Reagecon
BIOPHARMA
Product Code. 11513122 Shop All Reagecon Products
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Product Code. 11513122
Brand: Reagecon MTR04025
Used to calibrate, control, verify, or validate cryoscopes.
  • Cryoscope Reference Standard, Cryoscope Reference Material
  • Used to calibrate, control, verify or validate cryoscopes.
  • Manufactured for use with all cryoscopes compliant to International Reference Standard ISO5764/IDF108 for the determination of freezing point in milk.
  • Prepared gravimetrically on a weight/weight basis.
  • Both solute and solvent are weighed on a balance calibrated by Reagecon engineers using OIML traceable weights.
  • Reagecon holds ISO 17025 accreditation for calibration of laboratory balances (INAB Ref:265C).
  • The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
  • The certified values of each standard are verified using a high performance Cryoscope calibrated with NIST traceable, ISO Guide 34 Certified Standards.
Specifications
Packaging HDPE Flip-Top Bottle
Concentration or Composition Notes 621 (−0.600°C) (621m°H)
Physical Form Liquid
For Use With (Equipment) Cryoscope
Quantity 250 mL