missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRX/CORD2 Antibody (4G11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00001406-M02
This item is not returnable.
View return policy
Description
CRX/CORD2 Monoclonal antibody specifically detects CRX/CORD2 in Human, Mouse, Bovine, Fish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ChIP assay (ChIP)
Specifications
| CRX/CORD2 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| cone-rod homeobox, CORD2, CRDcone-rod homeobox protein, LCA7orthodenticle homeobox 3, OTX3 | |
| CRX (NP_000545, 1 a.a. ~ 95 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MMAYMNPGPHYSVNALALSGPSVDLMHQAVPYPSAPRKQRRERTTFTRSQLEELEALFAKTQYPDVYAREEVALKINLPESRVQVWFKNRRAKCR | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Bovine, Fish | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ChIP Assay | |
| 4G11 | |
| Western Blot 1:500, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP) | |
| NP_000545 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 1406 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction