missing translation for 'onlineSavingsMsg'
Learn More

CRISPR-Cas9 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-66379H

Product Code. 18289514

  • 625.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CRISPR-Cas9 Polyclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CRISPR-Cas9
Polyclonal
Western Blot
Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Full length S. pyogenes Cas9 protein [UniProt Q99ZW2]
0.1 mL
Primary
Bacteria
IgG
Western Blot
HRP
PBS with No Preservative
Rabbit
Affinity Purified
Gene Editing, Gene Knockouts, Gene Therapy
Cas9 protein from Streptococcus pyogene serotype M1.
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.