missing translation for 'onlineSavingsMsg'
Learn More

CRIF1 Antibody, Novus Biologicals™

Product Code. 18298252 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
25 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18298252 100 μL 100µL
18674717 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18298252 Supplier Novus Biologicals Supplier No. NBP258099

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CRIF1 Polyclonal specifically detects CRIF1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRIF1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias CKBBP2, CKII beta-associating protein, CR6 interacting factor 1, CRIF1p53-responsive gene 6 protein, growth arrest and DNA damage-inducible proteins-interacting protein 1, growth arrest and DNA-damage-inducible, gamma interacting protein 1, MGC4667, MGC4758, papillomavirus L2 interacting nuclear protein 1, Papillomavirus L2-interacting nuclear protein 1, Plinp1, PLINP-1CKII beta binding protein 2, PRG6CR6-interacting factor 1
Gene Symbols GADD45GIP1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ELEAEEREWYPSLATMQESLRVKQLAEEQKRRER
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 90480
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.