Learn More
Cresyl Violet Acetate High Purity Biological Stain MP Biomedicals
Product Code. 11432112 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432112 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11432112 Supplier MP Biomedicals Supplier No. 0215072705
  • ONLINE EXCLUSIVE PRICE
    122.00€
    115.50€ / 5g
    Save 6.50€
    5% Off
 Restricted Item
Estimated Shipment: 21-12-2026
High- purity biological stain used for staining nerve tissue, nerve cells, and fresh tumor tissue for biopsy.
Specifications
CAS 10510-54-0
Assay Percent Range ∽ 75%
Molecular Formula C18H15N3O3
MDL Number MFCD00013151,MFCD00013151
Synonym Cresyl Fast Violet, 9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt
Color Dark Green
Quantity 5 g
InChI Key XKOCOMKJPWEOHX-UHFFFAOYSA-M
SMILES CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
IUPAC Name acetic acid;5-iminobenzo[a]phenoxazin-9-amine
Molecular Weight (g/mol) 321.34
PubChem CID 44134641
Formula Weight 321.3
Grade Certified Biological Stain
Chemical Name or Material Cresyl Violet Acetate, Used for staining nerve tissue and nerve cells and fresh tumor tissue for biopsy.