missing translation for 'onlineSavingsMsg'
Learn More

Creatine Kinase BB Antibody, Novus Biologicals™

Product Code. 18058343 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18058343 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18058343 Supplier Novus Biologicals Supplier No. NB1002847

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Creatine Kinase BB Polyclonal specifically detects Creatine Kinase BB in Human samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Creatine Kinase B
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001814.2
Gene Alias Creatine Kinase Isoenzyme
Gene Symbols CKB
Host Species Goat
Immunogen Peptide with sequence C-KPSDEHKTDLNPDN corresponding to internal region according to NP_001814.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1152
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig, Cow .
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.