missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase BB Antibody (CKBB/6565), Novus Biologicals™
Mouse Monoclonal Antibody
249.00€ - 494.00€
Specifications
| Antigen | Creatine Kinase BB |
|---|---|
| Clone | CKBB/6565 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692105
|
Novus Biologicals
NBP3-20318-20ug |
20 μg |
249.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676904
|
Novus Biologicals
NBP3-20318-100ug |
100 μg |
494.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)Specifications
| Creatine Kinase BB | |
| 0.2 mg/ml | |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10mM PBS with 0.05% BSA | |
| 1152 | |
| IgG1 κ | |
| Protein A or G purified |
| CKBB/6565 | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Creatine Kinase Isoenzyme | |
| Recombinant human full-length Creatine Kinase BB protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title