missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase BB Antibody (8E4), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59462AF350
This item is not returnable.
View return policy
Description
Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| Creatine Kinase BB | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Creatine Kinase Isoenzyme | |
| Recombinant human Creatine Kinase BB (1-381aa) purified from E. coli | |
| 0.1 mL | |
| Cancer | |
| 1152 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 80000 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction