missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRBN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 12 publications
362.00€
Specifications
| Antigen | CRBN |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Simple Western 1:60, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Knockout Validated Validation (PMID: 31371704)., Single Cell Western, Knockdown Validated |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CRBN Polyclonal specifically detects CRBN in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated, Single Cell Western, Knockdown Validated.Specifications
| CRBN | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay, KnockDown | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| cereblon, DKFZp781K0715, MGC27358, MRT2A, non-syndromic, autosomal recessive, 2A, protein cereblon, protein x 0001 | |
| CRBN | |
| IgG | |
| Affinity Purified | |
| Specificity of human CRBN antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 μg/mL, Simple Western 1:60, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Knockout Validated Validation (PMID: 31371704)., Single Cell Western, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q96SW2 | |
| 51185 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EVEDQDSKEAKKPNIINFDTSLPTSHTYLGADMEEFHGRTLHDDDSCQVIPVLPQVMMILIPGQTLPLQLFHPQEVSMVRNLIQKDRTFAVLAYSNVQERE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title