missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRACC Monoclonal Antibody (520911)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561462
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
SLAMF7 contains one Ig-like C2-type (immunoglobulin-like) domain. Isoform 1 mediates NK cell activation through a SAP-independent extracellular signal-regulated ERK-mediated pathway. It may play a role in lymphocyte adhesion. Isoform 3 does not mediate any activation. SAP can bind the cytoplasmic tail of isoform 1 when phosphorylated in the presence of Fyn (in vitro). SLAMF7 is expressed in spleen, lymph node, peripheral blood leukocytes, bone marrow, small intestine, stomach, appendix, lung and trachea. Expression was detected in NK cells, activated B-cells, NK-cell line but not in promyelocytic, B-, or T-cell lines. The isoform 3 is expressed at much lower level than isoform 1. There are three named isoforms.
Specifications
| CRACC | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8BHK6 | |
| SLAMF7 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CRACC/SLAMF7, Ala22-Gly224. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 520911 | |
| Unconjugated | |
| SLAMF7 | |
| 19A; 19A24; 19A24 protein; 4930560D03Rik; CD2 subset 1; CD2-like receptor activating cytotoxic cells; CD2-like receptor-activating cytotoxic cells; CD319; CRACC; CS1; Leukocyte cell-surface antigen; membrane protein FOAP-12; Novel Ly9; novel LY9 (lymphocyte antigen 9) like protein; protein 19A; RGD1560190; SLAM family member 7; SLAMF7; UNQ576/PRO1138 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 75345 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction