missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRABP2 Mouse, anti-Human, CoraLite Plus 488, Clone: 1A5F3, Proteintech
Shop All Proteintech ProductsDescription
The CRABP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CRABP2. This antibody recognizes human, mouse, rat, pig antigen. The CRABP2 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
| Antigen | CRABP2 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Clone | 1A5F3 |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite Plus 488 |
| Gene Symbols | CRABP II, CRABP2, RBP6 |
| Host Species | Mouse |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 1382 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?