missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 550.00€
Specifications
| Antigen | CPSF2 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229647
|
Novus Biologicals
NBP3-38603-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232706
|
Novus Biologicals
NBP3-38603-20ul |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPSF2 Polyclonal antibody specifically detects CPSF2 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| CPSF2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| cleavage and polyadenylation specific factor 2, 100kDa, Cleavage and polyadenylation specificity factor 100 kDa subunit, cleavage and polyadenylation specificity factor subunit 2, CPSF 100 kDa subunit, CPSF 100kDa subunit, CPSF100, KIAA1367cleavage and polyadenylation specific factor 2, 100kD subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-700 of human CPSF2 (NP_059133.1).,, Sequence:, KGSFFKQAKKSYPMFPAPEERIKWDEYGEIIKPEDFLVPELQATEEEKSKLESGLTNGDEPMDQDLSDVPTKCISTTESIEIKARVTYIDYEGRSDGDSIKKIINQMKPRQLIIVHGPPEASQDLAECCRAFGGKDIKVYMPKLHETVDATSETHIYQVRLKDSLVSSLQFCKAKDAELAWIDGVLDMRVSKVDTGVILEEGELKDDGEDSEMQVEAPSDSSVIAQQKAMKSLFGDDEKETGEESEIIPTL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 53981 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title