missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam CPM, Thiol-Reactive Fluorescent Probe, >95% Purity, 50 mg
Description
- Molecular Weight: 402.44 g/mol
- Purity: >95%
- Properties: Widely used blue fluorescent thiol-reactive dye; non-fluorescent until reacted with thiols, allowing quantification without separation steps; acts as an energy acceptor from tryptophan and an energy donor to fluorescein.
- Applications: Monitors thiol release and distinguishes proliferating cancer cells via nucleolar protein staining.
Specifications
Specifications
| Product Type | Protein Labeling Reagent |
| Buffer | 7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin |
| Content And Storage | Store at -20°C. |
| Detection Location | In-Solution Detection |
| Label or Dye | Fluorescent Probe |
| Reactivity | Thiols |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | Thiols |
| Target Molecule | Thiols |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?