Learn More
Cole-Parmer™ CPC™ Quick-Disconnect Straight Coupling Insert
Product Code. 15437103
Click to view available options
Overall Length (Metric):
3.43 cm
3.76 cm
Unit Size:
Each
Product Code. 15437103
Brand: Cole-Parmer™ 3130422
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Quick-disconnect fitting with simple push-to-connect interface for your rigid tubing
  • Plug insert into body to connect fitting.
  • Fitting halves lock in place immediately.
  • To disconnect fitting halves, simply press the thumb latch on the coupling body.
  • spring-operated valve in each fitting half seals the flow path.
  • Straight-through fittings without shutoff valves are also available.
  • All fitting bodies have 316 stainless steel springs and latches.
Specifications
Type Quick-Disconnect Coupling Insert
Color White
Temperature (English) Max. 180°F
Temperature (Metric) Max 82°C
Port Type Quick-Disconnect x Push-to-Connect
Description 1/4 x 3/8 in.
Sterilization Method Autoclave or Ethylene Oxide (ETO)
Overall Length (Metric) 3.76 cm
Material Acetal
Pressure 120 psi