missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPA6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92859-0.1ml
This item is not returnable.
View return policy
Description
CPA6 Polyclonal antibody specifically detects CPA6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CPA6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| carboxypeptidase A6, CPAHCarboxypeptidase B, EC 3.4.17, EC 3.4.17.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-437 of human CPA6 (NP_065094.3). SLSGYNYEVYHSLEEIQNWMHHLNKTHSGLIHMFSIGRSYEGRSLFILKLGRRSRLKRAVWIDCGIHAREWIGPAFCQWFVKEALLTYKSDPAMRKMLNHLYFYIMPVFNVDGYHFSWTNDRFWRKTRSRNSRFRCRGVDANRNWKVKWCDEGASMHPCDDTYCGPFPESEPEVKAVANFLRKHRKHIRAYLSFHAYAQMLLYPYSYKYATIPNFRCVESAAYKAVNALQSVYGVRYRYGPASTTLYVSSGSSMDWAYKNGIPYAFAFELRDTGYFGFLLPEMLIKPTCTETMLAVKNITMHLLKKCP | |
| 0.1 mL | |
| Cancer, Cell Biology, Cytoskeleton Markers, Endocrinology, Neuroscience, Signal Transduction | |
| 57094 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction