missing translation for 'onlineSavingsMsg'
Learn More

COX7A1 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18631621 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18631621 0.02 mL 0.02mL
18603710 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18631621 Supplier Novus Biologicals Supplier No. NBP2921850.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COX7A1 Polyclonal antibody specifically detects COX7A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX7A1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias COX7AHCOX7A, COX7AM, cytochrome c oxidase subunit VIIa heart/muscle isoform, cytochrome c oxidase subunit VIIa polypeptide 1 (muscle), Cytochrome c oxidase subunit VIIa-H, Cytochrome c oxidase subunit VIIa-M, Cytochrome c oxidase subunit VIIa-muscle, mitochondrial
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-79 of human COX7A1 (NP_001855.1). MQALRVSQALIRSFSSTARNRFQNRVREKQKLFQEDNDIPLYLKGGIVDNILYRVTMTLCLGGTVYSLYSLGWASFPRN
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1346
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.