missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ COX4I1 (Human) Recombinant Protein

Human COX4I1 full-length ORF ( AAH08704, 1 a.a. - 169 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00001327-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16103421

  • 528.00€ / 25µg

Estimated Shipment: 11-07-2023
to see stock.
Add to basket

Description

Description

  • Sequence: MLATRVFSLVGKRAISTSVCVRAHESVVKSEDFSLPAYMDRRDHPLPEVAHVKHLSASQKALKEKEKASWSSLSMDEKVELYRIKFKESFAEMNRGSNEWKTVVGGAMFFIGFTALVIMWQKHYVYGPLPQSFDKEWVAKQTKRMLDMKVNPIQGLASKWDYEKNEWKK
Specifications

Specifications

AAH08704
cytochrome c oxidase subunit IV isoform 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COX4I1
GST
1327
Wheat germ expression system
25 μg
COX4, COXIV, MGC72016
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers