missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COTL1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149413
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes one of the numerous actin-binding proteins which regulate the actin cytoskeleton. This protein binds F-actin, and also interacts with 5-lipoxygenase, which is the first committed enzyme in leukotriene biosynthesis. Although this gene has been reported to map to chromosome 17 in the Smith-Magenis syndrome region, the best alignments for this gene are to chromosome 16. The Smith-Magenis syndrome region is the site of two related pseudogenes.
Specifications
| COTL1 | |
| Polyclonal | |
| Unconjugated | |
| Cotl1 | |
| 1810074P22Rik; 2010004C08Rik; CLP; coactosin like F-actin binding protein 1; coactosin-like 1; coactosin-like 1 (Dictyostelium); coactosin-like F-actin binding protein 1; coactosin-like protein; coactosin-like protein {ECO:0000250; Cotl1; cotl1 {ECO:0000312; EMBL:AAI58747.1, ECO:0000312; FLJ43657; MGC137989 protein; MGC19733; RGD:1305498}; UniProtKB:Q14019}; Unknown (protein for MGC:63983); wu:fb08h08; zgc:63983 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 23406, 361422, 72042 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| B0BNA5, Q14019, Q9CQI6 | |
| Cotl1 | |
| E. coli-derived recombinant human Coactosin-like Protein 1/COTL1, Ala2-Glu142. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction