missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Reusable Plastic Reagent Bottles with GL-45 PP Screw Cap

Product Code. 17293484 Shop All Corning Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
Unit Size:
Pack of 10
Pack of 20
Pack of 6
This item is not returnable. View return policy

Product Code. 17293484

Brand: Corning™ 1500P500

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Translucent polypropylene; with polypropylene screw cap.

These bottles are manufactured from translucent polypropylene. They are wide-mouth and supplied with PP screw top caps.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Color Translucent
Material Polypropylene
No. per Case 10/Cs.
No. per Pack 1/Pk.
Sterility Non-sterile
Shape Cylindrical
Height (Metric) 17.2 cm
Diameter (Metric) 8.7 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.